Rabbit Anti-IgG Polyclonal Antibody

ABCAAB6694-1EA 286 CHF
ABCAAB6694-1
Rabbit Anti-IgG Polyclonal Antibody
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR