Anti-GLB1 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)

ABCAAB6646-1EA 672 CHF
ABCAAB6646-1
Anti-GLB1 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)
Type: Primary
Antigen: GLB1 (galactosidase, beta 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR