Anti-Fluorescein Goat polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB6644-100EA 524.65 CHF
ABCAAB6644-100
Anti-Fluorescein Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Fluorescein (fluorescein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now