Anti-IgG Goat Polyclonal Antibody (Cy5®)

Supplier: Abcam
ABCAAB6564-500EA 280.12 CHF
ABCAAB6564-500
Anti-IgG Goat Polyclonal Antibody (Cy5®)
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR