Anti-GFP Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB6556-25EA 629.58 CHF
ABCAAB6556-25
Anti-GFP Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now