Anti-CDK9 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB6544-100EA 508.36 CHF
ABCAAB6544-100
Anti-CDK9 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CDK9 (cyclin-dependent kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR