Anti-IgG Donkey Polyclonal Antibody

Lieferant: Abcam
ABCAAB6520-1EA 0 CHF
ABCAAB6520-1
Anti-IgG Donkey Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Donkey
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR