Anti-HSV1 gG Envelope Protein Mouse Monoclonal Antibody [clone: 7F5]

Lieferant: Abcam
ABCAAB6511-100EA 486.49 CHF
ABCAAB6511-100
Anti-HSV1 gG Envelope Protein Mouse Monoclonal Antibody [clone: 7F5]
Antikörper
Type: Primary
Antigen: HSV1 gG Envelope Protein
Clonality: monoclonal
Clone: 7F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR