Anti-HSV1 gE Envelope Protein Mouse Monoclonal Antibody [clone: 9H3]

Supplier: Abcam
ABCAAB6510-100EA 456.49 CHF
ABCAAB6510-100
Anti-HSV1 gE Envelope Protein Mouse Monoclonal Antibody [clone: 9H3]
Antibodies
Type: Primary
Antigen: HSV1 gE Envelope Protein
Clonality: monoclonal
Clone: 9H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR