Anti-Cytomegalovirus pp65 Mouse monoclonal antibody unconjugated [clone: 3A12]

Supplier: Abcam
ABCAAB6503-100EA 741.79 CHF
ABCAAB6503-100
Anti-Cytomegalovirus pp65 Mouse monoclonal antibody unconjugated [clone: 3A12]
Antibodies
Type: Primary
Antigen: Cytomegalovirus pp65
Clonality: monoclonal
Clone: 3A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR