Anti-SLC8A1 Mouse monoclonal antibody [clone: 6H2]

ABCAAB6495-50EA 725 CHF
ABCAAB6495-50
Anti-SLC8A1 Mouse monoclonal antibody [clone: 6H2]
Type: Primary
Antigen: SLC8A1 (solute carrier family 8 (sodium/calcium exchanger), member 1)
Clonality: monoclonal
Clone: 6H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR