Anti-CDK2 Mouse monoclonal antibody unconjugated [clone: AN4.3]

Supplier: Abcam
ABCAAB6433-50EA 529.11 CHF
ABCAAB6433-50
Anti-CDK2 Mouse monoclonal antibody unconjugated [clone: AN4.3]
Antibodies
Type: Primary
Antigen: CDK2 (cyclin-dependent kinase 2)
Clonality: monoclonal
Clone: AN4.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR