Anti-Serotonin Rat Monoclonal Antibody [clone: YC5/45]
Lieferant: Abcam
ABCAAB6336-100EA 472.19 CHF
ABCAAB6336-100
Anti-Serotonin Rat Monoclonal Antibody [clone: YC5/45]
Antikörper
Type: Primary
Antigen: Serotonin
Clonality: monoclonal
Clone: YC5/45
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse