Anti-beta COP Mouse monoclonal antibody [clone: M3A5]

ABCAAB6324-100EA 708 CHF
ABCAAB6324-100
Anti-beta COP Mouse monoclonal antibody [clone: M3A5]
Type: Primary
Antigen: beta COP
Clonality: monoclonal
Clone: M3A5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR