Anti-NES Mouse Monoclonal Antibody [clone: 3k1]
Lieferant: Abcam
ABCAAB6320-50EA 634.35 CHF
ABCAAB6320-50
Anti-NES Mouse Monoclonal Antibody [clone: 3k1]
Antikörper
Type: Primary
Antigen: NES (nestin)
Clonality: monoclonal
Clone: 3k1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human