Anti-COL1 Mouse Monoclonal Antibody [clone: COL-1]

Supplier: Abcam
ABCAAB6308-50EA 596.55 CHF
ABCAAB6308-50
Anti-COL1 Mouse Monoclonal Antibody [clone: COL-1]
Antibodies
Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: monoclonal
Clone: COL-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR