Anti-CTNNB1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB6302-100EA 591.42 CHF
ABCAAB6302-100
Anti-CTNNB1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR