Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)

Lieferant: Abcam
ABCAAB6247-500EA 0 CHF
ABCAAB6247-500
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR