Anti-Fluorescein Mouse Monoclonal Antibody (Biotin) [clone: BT.6A4]

Supplier: Abcam
ABCAAB6215-200EA 492.8 CHF
ABCAAB6215-200
Anti-Fluorescein Mouse Monoclonal Antibody (Biotin) [clone: BT.6A4]
Antibodies
Type: Primary
Antigen: Fluorescein (fluorescein)
Clonality: monoclonal
Clone: BT.6A4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR