Anti-BDNF Rabbit polyclonal antibody

ABCAAB6201-250EA 732 CHF
ABCAAB6201-250
Anti-BDNF Rabbit polyclonal antibody
Type: Primary
Antigen: BDNF (brain-derived neurotrophic factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR