Anti-NGF Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB6199-250EA 430.55 CHF
ABCAAB6199-250
Anti-NGF Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NGF (nerve growth factor (beta polypeptide))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR