Anti-VIP Sheep Polyclonal Antibody

Supplier: Abcam
ABCAAB6184-100EA 903 CHF
ABCAAB6184-100
Anti-VIP Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VIP (vasoactive intestinal peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR