Anti-p75 NGF Receptor Mouse Monoclonal Antibody [clone: MC-192]

Supplier: Abcam
ABCAAB6172-100EA 456.49 CHF
ABCAAB6172-100
Anti-p75 NGF Receptor Mouse Monoclonal Antibody [clone: MC-192]
Antibodies
Type: Primary
Antigen: p75 NGF Receptor
Clonality: monoclonal
Clone: MC-192
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR