Anti-SNCB Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB6165-100EA 518.74 CHF
ABCAAB6165-100
Anti-SNCB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SNCB (synuclein, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR