Anti-IgG Goat Polyclonal Antibody (Texas Red®)

Supplier: Abcam
ABCAAB6109-500EA 259.37 CHF
ABCAAB6109-500
Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR