Anti-IgG Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB6107-500EA 207.49 CHF
ABCAAB6107-500
Anti-IgG Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR