Anti-GEMIN2 Mouse monoclonal antibody unconjugated [clone: 2E17]

Supplier: Abcam
ABCAAB6084-100EA 518.74 CHF
ABCAAB6084-100
Anti-GEMIN2 Mouse monoclonal antibody unconjugated [clone: 2E17]
Antibodies
Type: Primary
Antigen: GEMIN2 (gem (nuclear organelle) associated protein 2)
Clonality: monoclonal
Clone: 200000000000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR