Anti-AKT1 Rabbit polyclonal antibody

ABCAAB6076-1EA 691 CHF
ABCAAB6076-1
Anti-AKT1 Rabbit polyclonal antibody
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR