Anti-Thyroxine Mouse monoclonal antibody [clone: XM212]

ABCAAB6068-100EA 672 CHF
ABCAAB6068-100
Anti-Thyroxine Mouse monoclonal antibody [clone: XM212]
Type: Primary
Antigen: Thyroxine
Clonality: monoclonal
Clone: XM212
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR