Anti-TUBB Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB6046-100EA 529.11 CHF
ABCAAB6046-100
Anti-TUBB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR