Anti-PDLIM4 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB6045-100EA 870 CHF
ABCAAB6045-100
Anti-PDLIM4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDLIM4 (PDZ and LIM domain 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR