Anti-PDZD2 Goat polyclonal antibody

ABCAAB6037-100EA 458 CHF
ABCAAB6037-100
Anti-PDZD2 Goat polyclonal antibody
Type: Primary
Antigen: PDZD2 (PDZ domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR