Anti-KPNA2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB6036-100EA 505.57 CHF
ABCAAB6036-100
Anti-KPNA2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KPNA2 (karyopherin alpha 2 (RAG cohort 1, importin alpha 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR