Anti-PERP Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5986-100EA 518.74 CHF
ABCAAB5986-100
Anti-PERP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PERP (PERP, TP53 apoptosis effector)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR