Anti-IL22 Receptor Alpha Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB5984-100EA 500.8 CHF
ABCAAB5984-100
Anti-IL22 Receptor Alpha Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IL22 Receptor Alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human