Anti-TNFRSF17 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5972-100EA 529.11 CHF
ABCAAB5972-100
Anti-TNFRSF17 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNFRSF17 (tumor necrosis factor receptor superfamily, member 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR