Anti-LIN7B Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB5967-100EA 860 CHF
ABCAAB5967-100
Anti-LIN7B Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LIN7B (lin-7 homolog B (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR