Anti-GIPC1 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB5951-100EA 518.74 CHF
ABCAAB5951-100
Anti-GIPC1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GIPC1 (GIPC PDZ domain containing family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR