Anti-IgM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Abcam
ABCAAB5930-250EA 257.56 CHF
ABCAAB5930-250
Anti-IgM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now