Anti-ANLN Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB5910-100EA 492.8 CHF
ABCAAB5910-100
Anti-ANLN Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ANLN (anillin, actin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR