Anti-SENP7 Goat polyclonal antibody

ABCAAB5906-100EA 435 CHF
ABCAAB5906-100
Anti-SENP7 Goat polyclonal antibody
Type: Primary
Antigen: SENP7 (SUMO1/sentrin specific peptidase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR