Anti-Retinoic Acid Receptor gamma 1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB5904-100EA 486.49 CHF
ABCAAB5904-100
Anti-Retinoic Acid Receptor gamma 1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Retinoic Acid Receptor gamma 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR