Anti-BAG3 Goat polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB5898-100EA 903 CHF
ABCAAB5898-100
Anti-BAG3 Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: BAG3 (BCL2-associated athanogene 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR