Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Abcam
ABCAAB5883-500EA 286.17 CHF
ABCAAB5883-500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now