Anti-CREB Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB5803-100EA 529.11 CHF
ABCAAB5803-100
Anti-CREB Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CREB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR