Anti-PRKCH Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB5798-50EA 524.65 CHF
ABCAAB5798-50
Anti-PRKCH Rabbit polyclonal antibody
Type: Primary
Antigen: PRKCH (protein kinase C, eta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR