Anti-VAMP3 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB5789-100EA 524.65 CHF
ABCAAB5789-100
Anti-VAMP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VAMP3 (vesicle-associated membrane protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human