Anti-BAX Mouse monoclonal antibody [clone: 6A7]

ABCAAB5714-50EA 749 CHF
ABCAAB5714-50
Anti-BAX Mouse monoclonal antibody [clone: 6A7]
Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: monoclonal
Clone: 6A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR