Anti-IGF1R Rabbit polyclonal antibody

ABCAAB5679-50EA 826 CHF
ABCAAB5679-50
Anti-IGF1R Rabbit polyclonal antibody
Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR