Anti-SAE1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB5645-100EA 911 CHF
ABCAAB5645-100
Anti-SAE1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAE1 (SUMO1 activating enzyme subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR